Would you like to log in again?
TDM/DOA, Stanbio Laboratory
Supplier: Stanbio Laboratory
2410-051 2400-060
10152-566EA 469.22 USD InStock
10152-566 10152-456
TDM/DOA, Stanbio Laboratory
Clinical Diagnostic Supplies Clinical Reagents and Kits
Designed to be utilized on automated or manual chemistry analyzers, these assays are STAT procedures taking only minutes to get results and can be run directly on serum specimens with no extraction required.
Certifications: CE marked, FDA cleared, Health Canada Licensed.
Ordering information: TDM/DOA are ready-to-use liquid reagents.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty