Would you like to log in again?
LiquiColor® and Liqui-UV® Tests, Stanbio Laboratory
Supplier: Stanbio Laboratory
LiquiColor® & Liqui-UV®
2910-430 2960-500 2900-500 2930-430 2970-430 2960-430 2940-500 2940-430 2920-430 2900-430 2930-500 2920-500
10152-524EA 177.11 USD InStock
10152-524 10152-512 10152-468 10152-526 10152-470 10152-472 10152-460 10152-462 10152-464 10152-552 10152-554 10152-466
LiquiColor® and Liqui-UV® Tests, Stanbio Laboratory
Clinical Diagnostic Supplies Clinical Reagents and Kits
Stanbio Laboratory offers a broad range of liquid-stable clinical chemistry reagents
  • Absorbency of 405 nm for LiquiColor® Tests (Rate) and LiquiColor® Tests enzymes and 340 nm for Liqui-UV® Tests (Rate) enzymes
Our line of LiquiColor® and Liqui-UV® reagents are designed for maximum stability, ease-of-use and are optimized for today’s chemistry analyzers.
Alkaline Phosphatase LiquiColor® Test (Rate) (30 ml) by P-nitrophenylphosphate methodology: R1 (4×25 ml) and R2 (4×5 ml) and linearity of 800 U/L
Alkaline Phosphatase LiquiColor® Test (Rate) (125 ml) by P-nitrophenylphosphate methodology: R1 (4×105 ml) and R2 (4×20 ml) and linearity of 800 U/L
alpha--Amylase LiquiColor® Test (Rate) by EPS-G7 substrate method: R (4×30 ml) and linearity of 2000 U/L
ALT/SGPT Liqui-UV® Test (Rate) (30 ml) by IFCC recommended procedure: R1 (4×25 ml) and R2 (4×5 ml) and linearity of 600 U/L
ALT/SGPT Liqui-UV® Test (Rate) (125 ml) by IFCC recommended procedure: R1 (4×105 ml) and R2 (4×20 ml) and linearity of 600 U/L
AST/SGOT Liqui-UV® Test (Rate) (30 ml) by IFCC recommended procedure: R1 (4×25 ml) and R2 (4×5 ml) and linearity of 600 U/L
AST/SGOT Liqui-UV® Test (Rate) (125 ml) by IFCC recommended procedure: R1 (4×105 ml) and R2 (4×20 ml) and linearity of 600 U/L
CK Liqui-UV® Test (Creatine Kinase/Rate) by Szasz modified reaction method: R1 (4×24 ml) and R2 (4×6 ml) and linearity of 1200 U/L
LDH Liqui-UV® Test (Rate) (30 ml) by Modified Wacker methodology: R1 (4×25 ml) and R2 (4×5 ml) and linearity of 800 U/L
LDH Liqui-UV® Test (Rate), (125 ml) by Modified Wacker methodology: R1 (4×105 ml) and R2 (4×20 ml) and and linearity of 800 U/L
Gamma-GT LiquiColor® Test (g-Glutamyl Transferase/Rate) (30 ml) by Modified Szasz methodology: R1 (4×25 ml) and R2 (4×5 ml) and linearity of 300 U/L
Gamma-GT LiquiColor® Test (g-Glutamyl Transferase/Rate) (125 ml) by Modified Szasz methodology: R1 (4×105 ml) and R2 (4×20 ml) and linearity of 300 U/L
Certifications: All Enzymes are FDA cleared. Some enzymes have additional CE certifications.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty