Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-IgG Goat Polyclonal Antibody (DyLight® 800)

Supplier: Immunoreagents
GTXMU-003-E800NHSX
10150-296EA 127.96 USD InStock
10150-296
Anti-IgG Goat Polyclonal Antibody (DyLight® 800)
Antibodies
The ImmunoReagents' DyLight® 800 conjugates uses a near-IR fluorphore that is invisible to the naked eye but increases the staining options when using infrared imaging systems. DyLight® 800 has spectral properties that are very similar to other near-IR dyes, including Alexa Fluor 790 and IRDye 800, and has high solubility for maximal incorporation of the dye into the biomolecule of interest. Excitation/Emission = 777nm / 794nm Emission Color = Near IR (Similar Dyes: Alexa Fluor 790, IRDye 800)

  • Origin: US

ImmunoReagents' antibodies are suitable for most immunoassay formats and purified, cross adsorbed and conjugated based on the final product specfications.

Products will be accompanied with a standard product CoA to include application, purity and cross reactivity profiles.

Certifications: (EC) 142/2011 approved by APHIS, Reference Number is NC-TEC-0002

Ordering information: Products can by customized based on specific assay requirements: contact your VWR representative for more information.

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: DyLight® 800
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty