Would you like to log in again?
Stir Bar Kit, Egg-Shaped, PTFE, Chemglass
Supplier: Chemglass
CG-2005-20
80062-154EA 507.68 USD InStock
80062-154
Stir Bar Kit, Egg-Shaped, PTFE, Chemglass
Stirrers Magnetic Stirring Bars
Egg-shaped stir bars coated with PTFE
Kit contains six each of 20x10mm bars, six each of 30x16mm bars, three each of 40x20mm bars, two each of 50x20mm bars, and one 64x20mm bar. Replacement stir bars are available separately.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty