Would you like to log in again?
Whatman™ Filter Holder, Whatman products (Cytiva)
Supplier: Whatman products (Cytiva)
Whatman™
10461300 10461300
10035-082PK 161.21 USD InStock
CA11013-436 10035-082
Whatman™ Filter Holder, Whatman products (Cytiva)
Filter Holders
Filter holder made of Polysulfone for sampling and flow filtration, ideal for filtration with peristaltic pumps and for gas sterilization.
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty