Would you like to log in again?
Whatman™ Ashless Filter Aids, Whatman products (Cytiva)
Supplier: Whatman products (Cytiva)
Whatman™
1703-050 1703-050
10035-286PK 116 USD InStock
CA11013-932 10035-286
Whatman™ Ashless Filter Aids, Whatman products (Cytiva)
Filters Filter Paper Quantitative Filter Paper
Ashless filter aids enhance filtration speed by coagulating precipitates or suspensions to form a thick retentive 'prefilter' layer on top of normal filter paper.
  • Easily dispersible
  • Powder is of a purity similar to that of ashless quantitative papers
  • Maximum ash content is 0.015%
Ordering information: Supplied with two ended scoop for measuring 0.50 or 2.5 g quantities.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty