Would you like to log in again?
Absorption Chromatography Column with Fritted Disc, Chemglass
Supplier: Chemglass
CG-1199-36 CG-1199-41 CG-1199-40 CG-1199-25 CG-1199-30 CG-1199-13 CG-1199-17 CG-1199-35 CG-1199-26 CG-1199-16 CG-1199-22 CG-1199-12 CG-1199-20 CG-1199-31
80067-056EA 418.73 USD InStock
80067-056 80067-088 80067-046 80067-074 80067-062 80067-040 80067-054 80067-064 80067-080 80067-072 80067-082 80067-090 80067-038 80067-048
Absorption Chromatography Column with Fritted Disc, Chemglass
Chromatography Columns
Absorption chromatography column is useful for rapid preparative separations.
  • Column have a coarseporosity disc sealed into the lower end of the column for packing support
  • Moderate resolution can be obtained with this inexpensive to setup and operate column
  • Column is constructed using medium wall tubing
The Column can be used with optional CG-1201 flow controller and CG-1200 reservoir. Lower end of the column has a round bottom. Connections between the column, reservoir and flow control adapter are spherical joints connected with CG-150 stainless steel clamps.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty