Would you like to log in again?
OneRack® Tube Racks
Supplier: Heathrow Scientific
OneRack®
HS27513B HS27512A HS27512C
10147-534CS 253.41 USD InStock
10147-534 10147-765 10147-481
OneRack® Tube Racks
Racks Tube Racks
Durable tube racks are molded in a single, continuous piece so no assembly is required and there are no detachable pieces.
  • Made out of fiberglass-reinforced polyoxymethylene (POM)
  • Have molded coordinates for tube orientation
  • Stable design for use in water baths
  • Flat labeling area for easier identification
  • Unique teardrop-shaped openings for better retention of tubes
  • Offers the highest chemical and heat resistance (–40 to 121°C), autoclavable
Tough and more highly resistant to acids, bases, solvents and heat.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty