Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Parafilm M® Laboratory Film, Stoelting
Supplier: Stoelting
51359
100229-550EA 255 USD InStock
100229-550
Parafilm M® Laboratory Film, Stoelting
Seals General Purpose Sealing Films
Parafilm M® is a semi-transparent and flexible thermoplastic
It molds and seals tightly around test tubes, flasks, and irregular surfaces. It is moisture-proof, impervious to aqueous solutions, and protects anhydrous materials. The Dispenser/Cutter accommodates either 2" or 4" wide rolls, and a quick downward push cleanly cuts at the desired length.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty