Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-MARCKSL1 Rabbit Polyclonal Antibody

Supplier: Proteintech
11422-1-AP-150UL
10081-610EA 453 USD InStock
10081-610
Anti-MARCKSL1 Rabbit Polyclonal Antibody
Antibodies
MARCKS-like protein 1 (MARCKSL1) is widely expressed in nervous tissue, it is also named MARCKS-like protein (MLP) or MARCKS-related protein (MRP) and belongs to the MARCKS family, which is a highly acidic myristoylated family of PKC substrates widely distributed in diverse cell types including macrophages. The protein has a calculated molecular weight of 20 kDa and an apparent molecular weight of 42-48 kDa due to anomalous migration on SDS gels or phosphorylation. Genetic disruption of MARCKSL1 results in neural tube closure defects, events that depend on coordinated control of actin functions, cell shape, and cell migration. MARCKSL1 are thought to regulate the actin cytoskeleton and thereby participate in major cellular responses such as phagocytosis, secretion, motility, mitogenesis, and membrane trafficking.

Western Blot: Human Brain Tissue, 1:200-1:2000; IHC: Human lung cancer, 1:20-1:200

Type: Primary
Antigen: MARCKSL1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty