Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-IYD Rabbit Polyclonal Antibody

Supplier: Proteintech
16667-1-AP-150UL
10089-062EA 453 USD InStock
10089-062
Anti-IYD Rabbit Polyclonal Antibody
Antibodies
IYD(Iodotyrosine dehalogenase 1) is also named as C6orf71, DEHAL1 and belongs to the nitroreductase family. It is a transmembrane protein that deiodinates mono-and di-iodotyrosines (MIT, DIT) and recycles iodine, a scarce element in the environment, for the efficient synthesis of thyroid hormone. This protein has 6 isoforms produced by alternative splicing.

Western Blot: Human Liver Tissue, 1:200-1:2000

Type: Primary
Antigen: IYD
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty