Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-NOP56 Rabbit Polyclonal Antibody

Supplier: Proteintech
18181-1-AP-150UL
10091-212EA 453 USD InStock
10091-212
Anti-NOP56 Rabbit Polyclonal Antibody
Antibodies
Box C/D RNA-protein complexes (RNPs) guide the 2'-O-methylation of nucleotides in eukaryotic ribosomal RNAs. The box C/D and C'/D' RNP subcomplexes are each assembled with three sRNP core proteins. The Nop56 core protein mediates crucial protein-protein interactions required for both sRNP assembly and the methyltransferase reaction by bridging the L7Ae and fibrillarin core proteins . NOP56 is a component of the box C/D small nucleolar ribonucleoprotein complexes that direct 2-prime-O-methylation of pre-ribosomal RNA (rRNA) during its maturation. It is predicted to function in an early to middle step in pre-rRNA processing .

Western Blot: HeLa Cells, 1:200-1:2000

Type: Primary
Antigen: NOP56
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty