Would you like to log in again?
Tacky® Roll Mops Hand Held 6-301, Liberty
Supplier: Liberty Industries
10-1131
21924-076EA 112.13 USD InStock
21924-076
Tacky® Roll Mops Hand Held 6-301, Liberty
Mops
The Hand Held Tacky® Roll Mop has a 9" handle with one refill installed on the handle and is used on work surfaces, table tops, and hard-to-reach areas
It removes dirt, dust, debris, and lint from a variety of materials including glass, plastic, metal, cloth, and similar problem surfaces. The top sheet peels off to expose a fresh, clean surface when needed. The Hand Held Tacky Roll Mop has perforations about every 14" for easy access to a fresh sheet. For refills simply pull off the old core and replace it with a new roll.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty