Would you like to log in again?
Simulaids® Adult Airway Management Trainer
Supplier: NASCO HEALTHCARE INC. SE
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sort all by:
101-501
470120-426EA 3088.55 USD InStock
470120-426
Simulaids® Adult Airway Management Trainer
Models Human Biology Models
Practice Under Difficult Situations
  • Practice basic airway management
  • Incorporate common difficulty in intubation like tongue edema
  • Stomach can be filled to induce vomiting
Detailed anatomy featured on the adult intubation trainer stands out. Get a clear picture of human anatomy from the sculpted alveolar sac, bronchial tree, and blood vessel structures of the cross sectioned and exposed flexible lungs, to the interior main bronchus and oral/nasal pharyngeal spaces. Combine these details with an airway complicated by breakout teeth, tongue edema, and laryngospasm, and this airway trainer rates hard. An anatomical stomach that swells with esophageal intubation, or excessive pressure with the BVM during rescue breathing, together with the vomiting capability of this manikin makes this an exceptional educational tool for multitasking.
Delivery with instructions for use, soft carry bag, lubricant, and replaceable lungs and stomach.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty