Would you like to log in again?
3B Scientific® Deluxe Osteoporosis Model
Supplier: AMERICAN 3B SCIENTIFIC SE
1000153
470118-958EA 102.8 USD InStock
470118-958
3B Scientific® Deluxe Osteoporosis Model
Models Human Biology Models
This osteoporosis model consists of 3 medially divided lumbar vertebrae with intervertebral discs. For anatomical comparison, the upper section shows healthy bone structure, the middle section osteoporotic bone structure and the lower section advanced osteoporotic bone structure with flattened plates, deformation and decreased bone mass.
  • Highly detailed
  • Multiple planes of view
  • 3 year manufacturers warranty
  • Includes 3B smart anatomy
For detailed anatomical study the vertebrae can be removed from the stand.
Accessories information: For information about 3B smart anatomy, please see the document attached on this page.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty