Would you like to log in again?
3B Scientific® Kidney Stone Model
Supplier: AMERICAN 3B SCIENTIFIC SE
1000316
470119-172EA 117.5 USD InStock
470119-172
3B Scientific® Kidney Stone Model
Models Human Biology Models
  • Interchangeable Inserts For Male And Female Systems
  • 3 Year Manufacturers Warranty
  • Includes 3B Smart Anatomy
The kidney stone model is a helpful tool to inform patients about kidney stones (nephrolithiasis) and urinary stones (urolithiasis). Shown in the kidney stone model is an opened right kidney in natural size. The renal calices, the renal pelvis and the ureter are opened as well on the anatomy model so that concretions or stones can be identified in the following typical positions within the kidney: renal pyramids, origin of the upper calix group, renal cortex, connecting tubule of the lower calix group, causing congestion of the minor calices (partially closed, partially opened) in the kidney. Four original color pictures on the base of the kidney stone model show various kidney stones.
Accessories information: For information about 3B Smart Anatomy, please see the document attached on this page.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty