Would you like to log in again?
3B Scientific® Childbirth Demonstration Pelvis
Supplier: AMERICAN 3B SCIENTIFIC SE
1000334
470119-192EA 647.35 USD InStock
470119-192
3B Scientific® Childbirth Demonstration Pelvis
Models Human Biology Models
Easy To See Presentation
  • Realistic model of the childbirth process
  • 3 year manufacturers
This childbirth model demonstrates the progress of the fetal head through the pelvis during birth. The childbirth demonstration pelvis simulator consists of a female pelvic skeleton with a movable symphysis, hip bone, sacrum, coccyx and 2 lumbar vertebrae articulated to accommodate passage of a fetal skull mounted on an omni positioning flexible gooseneck support.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty