Would you like to log in again?
Life/form® Male Bladder Catheterization Simulator
Supplier: NASCO HEALTHCARE INC. SE
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sort all by:
LF00855
470119-368EA 1314 USD InStock
470119-368
Life/form® Male Bladder Catheterization Simulator
Models Human Biology Models
Allows Realistic Demonstration And Practice
  • Practice catheterization with a natural feel
  • Teaches students to safely and comfortably insert and remove a catheter
  • Includes carrying case
Catheterization is one of the basic skills all nursing students must learn, and to learn the techniques, they must practice. Any patient volunteers? Not likely! But the Life/form® simulator allows demonstration and practice in urinary catheterization without embarrassment or discomfort to the patient or the students. In addition, the catheterization simulator is useful for anatomical identification and demonstration of perineal care. With the re-designed Life/form® Male Catheterization Simulator, catheterization can be practiced just as on a real patient. A lubricated catheter can be inserted in the urethral orifice, passed through the urethra, and into the bladder. When the bladder is successfully entered, artificial urine (water) will flow from the catheter. The student will feel the normal restrictions caused by the mucosal folds, bulbous urethra, and the internal urethral sphincter, just prior to entrance into the bladder. The experience teaches proper positioning and movement of the penis to allow the catheter to pass easily with a minimum of discomfort to the patient. Just as in a real patient, the cuff must be completely deflated before the catheter is removed. Improper use of a Foley catheter can result in damage to the simulator, just as when using a Foley catheter with a real patient. Includes a hard carry case, lubricant, silicone catheter, and instruction manual.
Ordering information: Fluid supply stand not included.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty