Would you like to log in again?
Eisco® Premium Female Human Torso with Muscles and Open Back, 19 Parts
Supplier: EISCO SCIENTIFIC LLC
AM0009
470135-908EA 2636.7 USD InStock
470135-908
Eisco® Premium Female Human Torso with Muscles and Open Back, 19 Parts
Models Human Biology Models
Life size adult female human torso torso featuring an open back for in depth anatomical studies.
  • 19 parts
  • Hand painted
  • Mounted on base
  • Includes numbered key card
This 35" highly detailed anatomical model features muscles and an open back. Half of this model demonstrated muscular anatomy. The following removable structures are included: (1) head, (1) half brain, (1) eyeball, (1) liver with gall bladder, (1) small/large intestine - 4 part, (1) stomach - 2 part, (1) left lung with rib cage, (1) right lung with rib cage, (1) heart - 2 part, (2) vertebra, (1) female reproductive organs with 1st trimester fetus - 2 part, (1) inferior vena cava, aorta, iliac veins, iliac arteries.
Perfect core anatomical model for any high school classroom or biology enthusiast. It features 19 removable parts. Base of model measures 14.75×9.5" (37.5×24 cm).
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty