Would you like to log in again?
Eisco® Human Heart and Lungs Model
Supplier: EISCO SCIENTIFIC LLC
AM0068C
470136-094EA 229.19 USD InStock
470136-094
Eisco® Human Heart and Lungs Model
Models Human Biology Models
Dissectible model with 5 removable parts for viewing internal anatomical structures.
  • 3-dimensional model of human lungs, heart, and larynx
  • Model features 5 removable parts
  • Flat mounted on base
  • Hand painted
  • Includes numbered key card
Premium quality anatomical model of human lungs, heart, and larynx. Model features five removable parts. Heart can be removed and separated into two parts, right side lung can be removed for a cross section view, and larynx can be removed and separated to view internal structures. Realistic textures make this model an excellent tool for educational study, as well as for patient demonstrations. Made of durable, sturdy, and washable plastisol polymer, this replica will withstand long term use and handling. Model features artisan hand painted details and numbers, and includes a key card for identifying structures.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty