Would you like to log in again?
Eisco® Effects of Hypertension Model
Supplier: EISCO SCIENTIFIC LLC
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sort all by:
Eisco®
AM0083
470136-144EA 218.25 USD InStock
470136-144
Eisco® Effects of Hypertension Model
Models Human Biology Models
Depicts the damaging effects of hypertension on different organs and arteries.
  • Heart and kidney each dissect into two parts
  • All structures are mounted vertically to a common rod with base
  • Includes an information sheet outlining the resulting pathology associated with each structure
  • Designed by medical professionals and educators
  • Custom molded using the highest quality nontoxic polymers and PVC
  • Hand-painted by skilled artists
This unique model displays the harmful effects of hypertension on the most susceptible organs. It consists of scaled down depictions of the brain, eye, two-part heart, two-part kidney, and enlarged artery. The organs are displayed vertically on a common rod in the correct order, with a cutaway view of an artery mounted to the base. All of the organs can be rotated or removed for closer viewing. Includes an instruction sheet which gives information on the diseases associated with each structure as a result of hypertension. Great tool for showing the negative impacts of high blood pressure.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty