Would you like to log in again?
Hand-Held I.U.D Trainer
Supplier: NASCO HEALTHCARE INC. SE
160-1912
470119-230EA 45.35 USD InStock
470119-230
Hand-Held I.U.D Trainer
Models Human Biology Models
Economic Model Of Iud Placement
  • Sized for easy storage
  • Durable plastic construction
Hand-held trainer which is a suitable aid for understanding correct positioning of I.U.D. (Intrauterine Device) in the uterus. Made of durable plastic, the trainer features a transparent cover which allows easy visualization of insertion and placement of I.U.D. (I.U.D. not included). 
Ordering information: I.U.D is not Included.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty