Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Rhenium 1% on alumina spheres

Supplier: Thermo Scientific Chemicals
Synonyms: BCMMethyl 2-benzimidazolecarbamateMethyl benzimidazol-2-ylcarbamate

047023-30 47023-18
AA47023-30EA 1356.99 USD InStock
AA47023-30 AA47023-18
Rhenium 1% on alumina spheres
Rhenium
on 3mm (0.12in) alumina spheres


Formula: C9H9N3O2
Storage Temperature: Ambient
MDL Number: MFCD00011195
CAS Number: 7440-15-5
EINECS: 231-124-5
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty