Would you like to log in again?
Radglo GM-10 Chartreuse
Supplier: Thermo Scientific Chemicals
H36123-36 H36123-0B H36123-22
AAH36123-0BEA 384.75 USD InStock
AAH36123-0B AAH36123-36 AAH36123-22
Radglo GM-10 Chartreuse
Radglo GM-10
Used as a chartreuse colored fluorescent pigment for applications like PVC plastisols and organosols, PVC calendering, extruded and moulded polyolefines, and mass coloration of rubber.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty