Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-Cyclopentyloxy-4-methoxybenzyl alcohol 97%

Supplier: Thermo Scientific Chemicals

H62441-MD H62441-06 H62441-03
AAH62441-MDEA 45.59 USD InStock
AAH62441-MD AAH62441-06 AAH62441-03
3-Cyclopentyloxy-4-methoxybenzyl alcohol 97%
3-Cyclopentyloxy-4-methoxybenzyl alcohol


Formula: C₁₃H₁₈O₃
MW: 222.284 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03844627
CAS Number: 133332-49-7
Order Now
Specification Test Results
Assay 97%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty