Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Amino-2-chloro-3-fluorobenzeneboronic acid pinacol ester 96%

Supplier: Thermo Scientific Chemicals
Warning

H62733-MD H62733-03
AAH62733-03EA 452.99 USD InStock
AAH62733-03 AAH62733-MD
4-Amino-2-chloro-3-fluorobenzeneboronic acid pinacol ester 96%
4-Amino-2-chloro-3-fluorobenzeneboronic acid pinacol ester


Formula: C₁₂H₁₆BClFNO₂
Storage Temperature: Ambient
MDL Number: MFCD16996254
CAS Number: 1154740-65-4
Order Now
Specification Test Results
Assay 96%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty