Synonyms:
ADIBO-NHS ester, with 4 carbon linker
J64128-MB
J64128-MC
10067-578EA
165.34
USD
InStock
10067-578
10067-580
Azadibenzocyclooctyne-NHS ester, with 4 carbon linker
Azadibenzocyclooctyne-NHS ester, with 4 carbon linker
Azadibenzocyclooctyne-NHS ester, with 4 carbon linker is used in various applications like protein-peptide conjugates, antibody-enzyme or antibody-drug conjugates, protein or peptide-oligonucleotide conjugates and surface modification.
- Off-white to grey solid
- Moisture sensitive
- Soluble in DMF, THF, DMSO and DCM
Used to introduce cyclooctyne moiety in to biomolecules and amine containing compounds.
Caution: Incompatible with strong oxidizng agents.