Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Micronomicin sulphate 590 µg/mg

Supplier: Thermo Scientific Chemicals

J66072-09
10067-054EA 122.75 USD InStock
10067-054
Micronomicin sulphate 590 µg/mg
Micronomicin sulphate
Micronomicin sulfate is an aminoglycoside antibiotic commonly used against gram negative bacteria.

White to pale yellow powder

Applications: An aminoglycoside antibiotic commonly used against gram negative bacteria

Freely soluble in water

Caution: Research Use only: Not intended for animal or human diagnostic or therapeutic use.


Formula: C₂₀H₄₁N₅O₇·H₂SO₄
Boiling Pt: 667 °C
Flash Pt: >110 °C(230 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00941451
CAS Number: 66803-19-8
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty