Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Herbimycin A ≥95%

Supplier: Thermo Scientific Chemicals

J66427-MCR
10067-460EA 975.88 USD InStock
10067-460
Herbimycin A ≥95%
Herbimycin A
Herbimycin A is a sparingly soluble benzoquinone anti-tumor agent which interacts with the stress induced chaperone protein Hsp90 (heat shock protein 90).

  • Light Sensitive
  • Hygroscopic

Yellow powder

Applications: A sparingly soluble benzoquinone anti-tumor agent which interacts with the stress induced chaperone protein Hsp90 (heat shock protein 90)

Soluble in DMSO


Formula: C₃₀H₄₂N₂O₉
Boiling Pt: 752 °C
Flash Pt: >110 °C (230 °F)
Storage Temperature: Freezer
MDL Number: MFCD00151702
CAS Number: 70563-58-5
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty