Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Flucloxacillin sodium

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Flucloxacillin, sodium salt

J66615-MC
10067-470EA 100.48 USD InStock
10067-470
Flucloxacillin sodium
Flucloxacillin sodium
Targets primarily the cell wall of susceptible Gram-positive bacteria including Staphylococcus aureus.


Formula: C₁₉H₁₇ClFN₃NaO₅S
MW: 476.859 g/mol
Boiling Pt: 677 °C
Melting Pt: 176…178 °C
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD01682108
CAS Number: 1847-24-1
EINECS: 217-428-0
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty