Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Cefoxitin sodium 92.7-97%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: MefoxinCefoxitin sodium salt

J66800-03
10067-282EA 191.74 USD InStock
10067-282
Cefoxitin sodium 92.7-97%
Cefoxitin sodium
A broad spectrum antibiotic


Formula: C₁₆H₁₆N₃NaO₇S₂
MW: 449.428 g/mol
Boiling Pt: 843 °C
Melting Pt: >160 °C
Flash Pt: >110 °C(230 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00079042
CAS Number: 33564-30-6
EINECS: 251-574-6
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty