Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Amphotericin B 250 mg/ml in distilled water, sterile filtered

Supplier: Thermo Scientific Chemicals
Synonyms: (1R,3S,5R,6R,9R,11R,15S,16R,17R,18S,19E,21E,23E,25E,27E,29E,31E,33R,35S,36R,37S)-33-(((2R,3S,4S,5S,6R)-4-Amino-3,5-dihydroxy-6-methyltetrahydro-2H-pyran-2-yl)oxy)-1,3,5,6,9,11,17,37-octahydroxy-15,16,18-trimethyl-13-oxo-14,39-dioxabicyclo[33.3.1]nonatriaconta-19,21,23,25,27,29,31-heptaene-36-carboxylic acid

J67049-AD
AAJ67049-ADEA 156.01 USD InStock
AAJ67049-AD
Amphotericin B 250 mg/ml in distilled water, sterile filtered
Amphotericin B
  • Liquid

CAS No.: 1397-89-3
Storage Temperature: -30°C to -10°C
Physical Form: Liquid
MDL No.: MFCD00877763

Caution: Research use only: Not intended for animal or human diagnostic or therapeutic use.


Formula: C₄₇H₇₃NO₁₇
MW: 924.08 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00877763
CAS Number: 1397-89-3
EINECS: 231-791-2
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty