Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Human Fibroblast Growth Factor 7

Supplier: Thermo Scientific
J67052-AME J67052-EXE
10067-304EA 248.55 USD InStock
10067-304 10067-302
Human Fibroblast Growth Factor 7
Proteins and Peptides
Fibroblast Growth Factor 7, human

Formula Weight: 18.9 kDa
Storage Temperature: -30°C to -10°C
Physical Form: Lyophilized powder
Appearance: White
MDL No.: MFCD00212245

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty