Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.

Carbenicillin disodium salt 50 mg/mL in distilled water, sterile filtered

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: 6-[(Carboxyphenylacetyl)-amino]-3,3-dimethyl-7-oxo-4-thia-1-aza-bicyclo[3.2.0]heptane-2-carboxylic Acid, 2Naα-Carboxybenzylpenicillin disodium salt

J67159-AD J67159-AE
AAJ67159-ADEA 474.26 USD InStock
AAJ67159-AD AAJ67159-AE
Carbenicillin disodium salt 50 mg/mL in distilled water, sterile filtered
Carbenicillin disodium salt
Carbenicillin disodium salt, 50 mg/ml in distilled water, sterile-filtered


Formula: C₁₇H₁₆N₂O₆S·2Na
MW: 422.363 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00077683
CAS Number: 4800-94-6
EINECS: 225-360-8
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty