Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Mouse Recombinant Fibroblast Growth Factor

Supplier: Thermo Scientific
J67243-EXE J67243-IAM
10067-374EA 126.9 USD InStock
10067-374 10067-372
Mouse Recombinant Fibroblast Growth Factor
Proteins and Peptides
Fibroblast Growth Factor, basic, mouse

Formula Weight: 16.2 kDa
Storage Temperature: -30°C to -10°C
Physical Form: Lyophilized powder
Appearance: White

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty