Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Rifampicin 50 mg/ml, sterile filtered

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: Rifamycin AMP3-(4-Methylpiperazinyliminomethyl)rifamycin SVRifampin

J67397-XF
AAJ67397-XFEA 164.87 USD InStock
AAJ67397-XF
Rifampicin 50 mg/ml, sterile filtered
Rifampicin
Liquid


Formula: C₄₃H₅₈N₄O₁₂
MW: 822.95 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00151389
CAS Number: 13292-46-1
EINECS: 236-312-0
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty