Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Human Recombinant Interleukin 4

Supplier: Thermo Scientific
J67406-YAM
10067-536EA 165.54 USD InStock
10067-536
Human Recombinant Interleukin 4
Proteins and Peptides
Interleukin 4, human

Formula Weight: 15 kDa
Storage Temperature: -30°C to -10°C
Physical Form: Lyophilized powder
Appearance: White
MDL No.: MFCD00151705

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty