Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Safranine O for Gram staining

Supplier: HARDY DIAGNOSTICS
Synonyms: Safranin TSafranin Y3,7-Diamino-2,8-dimethyl-5-phenylphenazinium chlorideSafranin OGossypimine3,7-Diamino-2,8-dimethyl-5-phenylphenazin-5-ium chlorideCI 50240Basic Red 2Cotton RedSafranin ASafranin

S008
89428-930EA 12.46 USD InStock
89428-930
Safranine O for Gram staining
Safranine O
Safranin is a traditional counterstain used in gram staining procedures. An advanced counterstain is also available for improved staining. Organisms stained with the advanced counterstain will appear a brighter red with better defined edges. Stained cells are less likely to be overwashed.


Formula: C₂₀H₁₉ClN₄
MW: 350.85 g/mol
MDL Number: MFCD00011759
CAS Number: 477-73-6
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty