Would you like to log in again?
Dosys™ Syringe Feed Tubing, DWK Life Sciences
Supplier: DWK Life Sciences
851419
53500-657EA 38.93 USD InStock
53500-657
Dosys™ Syringe Feed Tubing, DWK Life Sciences
Tubing Transfer Tubing
These tubing is made of long lasting medical quality silicone.
  • Available in various length, cut or uncut
  • Autoclavable at 121 °C
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty