Would you like to log in again?
Trypsin Inhibitor. Powder
Supplier: MP BIOMEDICALS (FKA ICN BIOMED
0210061201 02100612.5 0210061205 02100612.2
IC100612.5EA 220.77 USD InStock
IC100612.5 IC10061205 IC100612.2 IC10061201
Trypsin Inhibitor. Powder
Proteins and Peptides
The ability of the inhibitor to inhibit the action of bovine trypsin is measured spectrophotometrically in a system in which benzoyl-arginine-ethyl ester is utilized as the typsin substrate.
Ovomucoid or trypsin inhibitor is an abundant protein in most avian egg whites. The hen′s egg protein is composed of about 186 amino acid residues, and is highly glycosylated.
A trypsin inhibitor prevents the trypsin enzyme from breaking down the protein or reduces the enzyme's efficiency.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty