Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Butyric acid ≥99.5%, liquid

Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Danger
Synonyms: n-Butyric acidButanoic acid

0210116980
IC10116980EA 53.3 USD InStock
IC10116980
Butyric acid ≥99.5%, liquid
Butyric acid
Butyric acid is used in the preparation of various butyrate esters. Low-molecular-weight esters of butyric acid, such as methyl butyrate, have mostly pleasant aromas or tastes. As a consequence, they find use as food and perfume additives. It is also used as an animal feed supplement, due to the ability to reduce pathogenic bacterial colonization.


Formula: C₄H₈O₂
MW: 88.11 g/mol
MDL Number: MFCD00002814
CAS Number: 107-92-6
UN: 2820
ADR: 8,III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty