Would you like to log in again?
Bovine Casein Hydrolysate Acid
Supplier: MP Biomedicals
0210129105 0210129125 0210129101
IC10129105EA 587.55 USD InStock
IC10129105 IC10129125 IC10129101
Bovine Casein Hydrolysate Acid
Proteins and Peptides
Soluble in water(100 mg/mL).
Product is prepared by acid digestion of casein.
Total Nitrogen : ~8.2% ; Amino Nitrogen ~6.1% ; Chlorides (as NaCl) ~35% ; Standard Plate Count <10,000/g
Casein, Acid Hydrolysate is a hydrochloric acid hydrolysate of casein for use in preparing microbiological culture media. Casein is a rich source of amino nitrogen. Treatment with hydrochloric acid hydrolyzes the protein into primarily free amino acids with some short peptide fragments. Tryptophan and vitamins present in the casein are destroyed by the acid hydrolysis. The salt content of this peptone is very high at approximately 36%.
Casein acid hydrolysate is used as a microbiological nutrient in laboratory media and fermentations. It has also been used as a nutrient in vaccine and toxin production where a high free amino acid content is required.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty