Would you like to log in again?
Horse Cytochrome C (from Heart)
Supplier: MP Biomedicals
02101467.2 02101467.1 0210146701 02101467.5
IC101467.1EA 312.54 USD InStock
IC101467.1 IC101467.2 IC101467.5 IC10146701
Horse Cytochrome C (from Heart)
Proteins and Peptides
Cytochrome c is an electron-carrying mitochondrial protein. The transition of Cytochrome c between the ferrous and ferric states within the cell makes it an efficient biological electron-transporter and it plays a vital role in cellular oxidations in both plants and animals. It is generally regarded as a universal catalyst of respiration, forming an essential electron-bridge between the respirable substrates and oxygen.
  • Identity test: Passes
Cytochrome c is generally regarded as a universal catalyst of respiration, forming an essential electron-bridge between the respirable substrates and oxygen.
Dark red to red-brown solution at 10 mg plus 1 ml of water.
Caution: Research Use Only (RUO).
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty