Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Maleic acid, white crystalline powder, Technical Grade

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

0210223201
IC10223201EA 122.57 USD InStock
IC10223201
Maleic acid, white crystalline powder, Technical Grade
Maleic acid
Maleic acid (MA) is a dicarboxylic acid that undergoes esterification with ethanol in the presence of cation-exchange resin catalysts to form diethyl maleate. Its crystalline structure has been analyzed. Reaction kinetics and mechanism of the isomerization of maleic acid to fumaric acid in the presence of ceric and bromide ions has been investigated. The reaction mechanism and reactivity ratios for the radical copolymerization of maleic acid with styrene have been determined.

Soluble in Water. (1% aq soln, 333 mg/mL).


Formula: C₄H₄O₄
MW: 116.07 g/mol
MDL Number: MFCD00063177
CAS Number: 110-16-7
UN: 3261
ADR: 8,III
https://us-legacy.vwr.com/stibo/web/std.lang.all/72/21/2297221.jpg
Order Now
Specification Test Results
Identity Test Passes
FTIR Conforms to standard
Moisture Content ≤5%
Melting Point 130 - 145°C
Solubility (1% aq.) Clear and colorless

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty