Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium selenite ∼98%, powder Cell Culture Reagent

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

0210292825
IC10292825EA 103.11 USD InStock
IC10292825
Sodium selenite ∼98%, powder Cell Culture Reagent
Sodium selenite
Sodium selenite is commonly used as a source of selenium in biological research, where selenium is an essential trace element that is normally provided by serum. Selenium is present in selenoproteins such as glutathione peroxidase and thioredoxin reductase, which contain the selenium analog of cysteine, selenocysteine. In particular, glutathione peroxidase has a role in detoxification in vivo as a scavenger of peroxides. Sodium selenite is included in various medium supplements for use in cell culture. Sodium selenite has been utilized in studies of cell proliferation and cancer research. It has been used to alter gene expression in HepG2 cells as analyzed by cDNA microarrays. Sodium selenite can inhibit zinc finger protein/DNA interactions. Sodium selenite has been shown to alter mitochondrial membrane potentials and thus potentially contribute to apoptosis.

More potent than selenomethionine in reducing carcinogenesis.

Selenium Content ≥44.0%

Soluble in water.

Sodium selenite is commonly used as a source of selenium in biological research, where selenium is an essential trace element that is normally provided by serum. Selenium is present in selenoproteins such as glutathione peroxidase and thioredoxin reductase, which contain the selenium analog of cysteine, selenocysteine. In particular, glutathione peroxidase has a role in detoxification in vivo as a scavenger of peroxides. Sodium selenite is included in various medium supplements for use in cell culture. Sodium selenite has been utilized in studies of cell proliferation and cancer research. It has been used to alter gene expression in HepG2 cells as analyzed by cDNA microarrays. Sodium selenite can inhibit zinc finger protein/DNA interactions. Sodium selenite has been shown to alter mitochondrial membrane potentials and thus potentially contribute to apoptosis.


Formula: Na₂SeO₃
MW: 172.94 g/mol
MDL Number: MFCD00003489
CAS Number: 10102-18-8
UN: 2630
ADR: 6.1,I
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty