Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ammonium carbonate, white powder

Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Synonyms: Salt of hartshorn

0215010880
IC15010880EA 68.35 USD InStock
IC15010880
Ammonium carbonate, white powder
Ammonium carbonate
Ammonium carbonate is used in the washing and defatting of wools, the manufacture of rubber products, in fire extinguishers, and the separation of cacao constituents.

A high-throughput parallel liquid chromatography/ultraviolet/mass spectrometry (LC/UV/MS) technique for compound screening that incorporates ammonium carbonate has been published. Ammonium carbonate can be used in the isolation of cell surface proteins from Candida albicans. It also finds application in the comparative peptide analysis of equine, bovine, and canine cytochrome c by capillary electrophoresis/mass spectrometry. The preparation of glycoconjugates via Kochetkov aminations in saturated aqueous ammonium carbonate has been reported.

Soluble in 4 parts water, decomposed by hot water.

Caution: Research Use Only (RUO).


Formula: (NH₄)₂CO₃
MW: 96.09 g/mol
Melting Pt: 58 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00010890
CAS Number: 10361-29-2
Order Now
Specification Test Results
Identity Test Passes

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty