Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Adenosine 5'-monophosphate disodium salt (AMP disodium salt) hydrate ≥99%, Powder

Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Synonyms: 5′-AMP-Na2 hydrate

0215026405
IC15026405EA 97.52 USD InStock
IC15026405
Adenosine 5'-monophosphate disodium salt (AMP disodium salt) hydrate ≥99%, Powder
Adenosine 5'-monophosphate disodium salt (AMP disodium salt) hydrate


Formula: C₁₀H₁₂N₅Na₂O₇P·H₂O
MW: 391.2 g/mol (anhy)
MDL Number: MFCD00065023
CAS Number: 4578-31-8
Order Now
Specification Test Results
Synonyms AMP; 5'-Adenylic acid; 5′-AMP-Na2
Form White Powder
Storage Store at -0 °C.
Optical Rotation [α]20/D -43 ± 2°, c = 1% in 0.5 M Na2HPO4 (dry matter) (Lit.)
pH 7 (Lit.)
Source Yeast

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty