Would you like to log in again?
Avidin (from Egg white)
Supplier: MP Biomedicals
0215040725 0215040710 0215040705 0215040701 0215040780
IC15040701EA 34.62 USD InStock
IC15040701 IC15040710 IC15040705 IC15040725 IC15040780
Avidin (from Egg white)
Proteins and Peptides
Soluble in water or dilute aqueous buffer.
One unit will bind 1 µg of D-biotin at pH 8.9.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty