Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Bromocresol purple sodium salt, orange powder

Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Warning
Synonyms: Sodium 2-bromo-4-(3-(3-bromo-4-hydroxy-5-methylphenyl)-1,1-dioxido-3H-benzo[c][1,2]oxathiol-3-yl)-6-methylphenolateSodium α-(3-bromo-5-methyl-4-oxo-2,5-cyclohexadienylidene)-α-(3-bromo-5-methyl-4-hydroxyphenyl)toluenesulphonateSodium α-(3-bromo-5-methyl-4-oxo-2,5-cyclohexadienylidene)-α-(3-bromo-5-methyl-4-hydroxyphenyl)toluenesulfonateBromocresol purple water soluble

0215051805
IC15051805EA 52.67 USD InStock
IC15051805
Bromocresol purple sodium salt, orange powder
Bromocresol purple sodium salt
Bromocresol purple sodium salt is typically used as a pH indicator (5.2 - 6.8 : yellow to blue-purple). Used in spectrophotometric determination of serum-albumin.

Soluble in water (80 mg/ml), ethanol (20 mg/ml) or methyl cellosolve (90 mg/ml).


Formula: C₂₁H₁₅Br₂NaO₅S
MW: 562.21 g/mol
Melting Pt: 255 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00148896
CAS Number: 62625-30-3
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty