Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Deoxy-D-galactose, white powder

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

0215080001
IC15080001EA 136.75 USD InStock
IC15080001
2-Deoxy-D-galactose, white powder
2-Deoxy-D-galactose
Inhibitor of fucosylation.

Caution: Research Use Only (RUO).


Formula: C₆H₁₂O₅
MW: 164.16 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00014649
CAS Number: 1949-89-9
Order Now
Specification Test Results
Identity Test Passes
Melting Point 109 - 119°C
FTIR Conforms to Standard
Optical Rotation (c=4, H₂O) +54.8° to +66.3°

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty